Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GERPN8512

Nucleon BACC3

Cytiva RPN8512, pack of 1 kit, sufficient for 50 preparations

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.55
Pricing and availability is not currently available.

usage

sufficient for 50 preparations

packaging

pack of 1 kit

manufacturer/tradename

Cytiva RPN8512

General description

Nucleon BACC system rapidly extracts high molecular weight DNA from whole blood and cultured cells. The Nucleon proprietary resin is added following cell lysis, deproteinization with sodium perchlorate, and a single chloroform extraction.

Application

Developed for rapid extraction of high-quality, high molecular weight genomic DNA from blood and cell cultures.

Features and Benefits

  • Developed for rapid extraction of high-quality, high molecular weight genomic DNA from blood and cell cultures.
  • Features a phenol-free protocol requiring only 30 min to complete.
  • For 50 preps from up to 10 mL of whole blood (kit contains all reagents necessary for 50 x 10 mL blood extractions).

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

pictograms

Exclamation mark

signalword

Danger

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service