Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GERPN4402

AMDEX Streptavidin Alkaline Phosphatase Conjugate

Cytiva RPN4402

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

form

liquid

packaging

pkg of 0.5 mL

manufacturer/tradename

Cytiva RPN4402

shipped in

wet ice

storage temp.

2-8°C

General description

Streptavidin - Alkaline Phosphatase. Streptavidin conjugated to multiple alkaline phosphatase molecules on a Dextran backbone.

Features and Benefits

  • It can be used in direct substitution for a standard anti-rabbit HRP conjugate, without the need to alter existing protocols.
  • It is important to optimize the dilution by setting up a range test.
  • Option to reduce incubation times.
  • Well-controlled nonspecific binding properties.

Storage and Stability

Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

AMDEX is a trademark of Cytiva

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service