Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GERPN2634

MMP-9 Biotrak activity asay

Cytiva RPN2634

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

manufacturer/tradename

Cytiva RPN2634

packaging

pkg of 96 wells

shipped in

dry ice

Storage temp.

−20°C

General description

Biotrak MMP-9, Activity Assay System Companies provides a simple, specific and precise quantitative determination of active or pro MMP-9 in tissue culture supernatants, serum, plasma, synovial fluid and tissue homogenates. Active MMP-9 may be measured in the range 0.5 to 16 ng/mL for normal endogenous MMP-9 levels. A longer incubation protocol enables an assay range of 0.125 to 4 ng/mL for lower endogenous MMP-9 levels. The assay uses the pro form of a detection enzyme that can be activated by captured active MMP-9, into an active detection enzyme, through a single proteolytic event. The natural activation sequence in the pro detection enzyme has been replaced using protein engineering, with an artificial sequence recognized by specific matrix metalloproteinases.

MMP activated detection enzyme can then be measured using a specific chromogenic peptide substrate. Standards and samples are incubated in microtitre wells precoated with anti-MMP-9 antibody. Any MMP-9 present will be bound to the wells, other components of the sample being removed by washing and aspiration. Either the endogenous levels of free active MMP-9, or the total levels of free MMP-9 in a sample, can be detected. In order to measure the total MMP-9 content, any bound MMP-9 in its pro form is activated using p-Aminophenylmercuric Acetate (APMA). The standard is pro MMP-9 which is activated in parallel for both types of sample. Active MMP- 9 is detected through activation of the modified pro detection enzyme and the subsequent cleavage of its chromogenic peptide substrate. The resultant colour is read at 405 nm in a microplate spectrophotometer. The concentration of active MMP-9 in a sample is determined by interpolation from a standard curve. Each pack contains reagents for 96 determinations. This allows the construction of a standard curve plus the measurement of 42 samples in duplicate. The Biotrak MMP-9 activity assay system has been specifically designed for research purposes.

Features and Benefits

  • Specific for MMP-9.
  • Measures endogenous active MMP-9 or total active MMP-9 (following activation using APMA).
  • Nonradioactive microplate-based assay.
  • Precise and accurate measurement.
  • Applicable to complex biological samples.
  • Ready-to-use detection enzyme and substrate.
  • Equivalent sensitivity to ELISA

Storage and Stability

Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Biotrak MMP-1, MMP-3, MMP-9, and TIMP-1 Human ELISAs
These products are developed in collaboration with FUJI CHEMICAL INDUSTRIES, Ltd.(Japan).
Biotrak is a trademark of Cytiva

pictograms

Environment

signalword

Danger

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service