Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GERPN2610

MMP-1, Human, Biotrak ELISA

Cytiva RPN2610, pack of 1 ea, size 96 wells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

packaging

pack of 1 ea

manufacturer/tradename

Cytiva RPN2610

size

96 wells

shipped in

dry ice

storage temp.

−20°C

General description

MMP-1, Human, Biotrak Assay.

Application

The MMP-1 Human Biotrak ELISA system has been specifically designed for research purposes. MMP-1 may be measured in the range 6.25-100 ng/mL.

The assay is based on a two site ELISA ‘sandwich′ format. Standards and samples are incubated in microplate wells pre-coated with anti-MMP-1 antibody. Any MMP-1 present will be.
bound to the wells, other components of the sample being removed by washing and aspiration. A second polyclonal antibody to MMP-1 is then bound to the MMP-1. Any excess is removed by washing and aspiration. The second antibody bound to the wells is detected using donkey anti-rabbit horseradish peroxidase. The amount of peroxidase bound to each well is determined by the addition of TMB ‘ready-to-use′ substrate. The reaction is stopped by addition of an acid solution, and the resultant color read at 450 nm in a microplate spectrophotometer. The concentration of MMP-1 in a sample is determined by interpolation from a standard curve.

Each pack contains reagent for 96 determinations. This allows the construction of a standard curve plus the measurement of 42 samples in duplicate.

Features and Benefits

  • Measures total MMP-1.
  • Specific for human MMP-1.
  • Precise and accurate measurement.
  • Non-isotopic protocol.
  • Ready to use substrate.
  • Same day protocol.

Storage and Stability

Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Biotrak is a trademark of Cytiva

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service