GERPN2255
cAMP Biotrak™ EIA, (Non-Acetylation Protocol, Lysis Reagents not included)
Cytiva RPN2255, pack of 5
About This Item
Recommended Products
packaging
pack of 5
manufacturer/tradename
Cytiva RPN2255
storage temp.
2-8°C
General description
The Biotrak™ cAMP competitive enzymeimmunoassay system is specifically designed for research purposes. It uses peroxidase-labeled cAMP conjugate, a specific antiserum which can be immobilized on to precoated microplates, and a one-pot stabilized substrate solution.
RPN2255 contains one vial (with 5× the volume of RPN225), for each of the following components:
Conjugate,
Antiserum,
Assay buffer,
Wash buffer,
Substrate
There are also five vials of non-acetylation standard, each containing the same volume as the component supplied in RPN225, and five coated plates.
Features and Benefits
- Enzyme immunoassay - Bulk non-acetylation kit.
- Range 25 - 6400 fmol/well (non-acetylation protocol).
- Rapid assay protocol.
- Nonradioactive.
- Specific for cAMP.
- Precise and accurate measurement
- Ready-to-use substrate
- Color-coded reagents.
Storage and Stability
Analysis Note
Legal Information
signalword
Warning
hcodes
Storage Class
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service