Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GERPN224

Leukotriene C4/D4/E4, Biotrak EIA System

Cytiva RPN224

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

manufacturer/tradename

Cytiva RPN224

packaging

pkg of 96 wells

shipped in

dry ice

storage temp.

−20°C

General description

Leukotriene C4/D4/E4 Biotrak enzymeimmunoassay system from Cytiva has been specifically designed for research purposes. It combines the use of a Peroxidase labelled Leukotriene C4 conjugate, a peptido Leukotriene specific antiserum which can be immobilized on to pre-coated microplates, and a one pot stabilized substrate solution. This provides a rapid and sensitive non-isotopic method for the determination of Leukotriene C4/D4/E4 in the range 0.77 pg to 49.5 pg/well. Each pack contains sufficient material for 96 wells. This permits the construction of one standard curve and 39 unknowns in duplicate. The assay is based on the competition between unlabelled Leukotriene C4 and a fixed quantity of Peroxidase labelled Leukotriene C4 for a limited number of binding sites on a peptido leukotriene specific antibody.

With fixed amounts of antibody and Peroxidase labelled Leukotriene C4 the amount of Peroxidase labelled ligand bound by the antibody will be inversely proportional to the concentration of added unlabelled ligand. The Peroxidase ligand that is bound to the antibody is immobilized on to polystyrene microplate wells precoated with second antibody, as demonstrated in figure 1. Thus any unbound ligand can be removed from the well by simple washing procedures. The amount of Peroxidase labelled Leukotriene C4 bound to the antibody is determined by addition of Tetramethylbenzidine (TMB)/ Hydrogen Peroxide single pot substrate. The reaction is stopped by addition of an acid solution, and the resultant colour read at 450 nm in a microplate spectrophotometer. The concentration of unlabelled Leukotriene C4 in a sample is determined by interpolation from a standard curve.

Features and Benefits

  • High sensitivity ~ 0.5 pg/well for Leukotriene C4/D4, 10 pg/mL.
  • Nonisotopic.
  • Ready-to-use substrate.
  • Color-coded reagents.
  • Choice of protocol.

Storage and Stability

Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Biotrak is a trademark of Cytiva

pictograms

Flame

signalword

Warning

hcodes

Storage Class

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service