GERPN2020N
Hybond®-N hybridization membranes
W × L 20 cm × 20 cm, sheets, pack of 10 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
material
neutral nylon
sheets
form
solid
packaging
pack of 10 ea
manufacturer/tradename
Cytiva RPN2020N
W × L
20 cm × 20 cm
color
White
pore size
0.45 μm pore size
Looking for similar products? Visit Product Comparison Guide
Legal Information
Hybond is a registered trademark of Cytiva
Storage Class
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service