GERPN1636
Rapid-hyb Buffer
Cytiva RPN1636, pack of 500 mL
About This Item
Recommended Products
sterility
non-sterile
packaging
pack of 500 mL
manufacturer/tradename
Cytiva RPN1636
storage condition
room temperature (Under these conditions the product is stable for at least 3 months)
General description
Storage and Stability
Analysis Note
related product
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service