Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GERPN1604

Megaprime DNA Labelling System, dNTP

Cytiva RPN1604, sufficient for 30 reactions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106000
NACRES:
NA.31

usage

sufficient for 30 reactions

manufacturer/tradename

Cytiva RPN1604

shipped in

dry ice

storage temp.

−20°C

General description

Megaprime DNA Labeling System, dNTP (for use with any radioactively labeled nucleotide).

Application

Megaprime DNA Labeling Systems are used for rapid, random-prime labeling from as little as 25 ng substrate DNA. Each system includes the following reagents, sufficient for 30 or 60 reactions (depending on product ordered): primer solution, labeling buffer, reaction buffer, enzyme, standard DNA solution, and carrier DNA solution.

Features and Benefits

  • For labeling of DNA probes to specific activities > 1 × 109 dpm/μg in 10 min.
  • Features cloned Klenow fragment and random nonamers in the random-prime labeling technique.
  • No need to remove unincorporated nucleotides before hybridization.
  • Protocols for labeling directly from Low melting point Agarose.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Megaprime is a trademark of Cytiva

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service