Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GERPN1510B

Hybond®-N+ hybridization membranes

W × L 15 cm × 10 cm, sheets, pack of 20 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104921
NACRES:
NB.22

material

sheets

packaging

pack of 20 ea

manufacturer/tradename

Cytiva RPN1510B

W × L

15 cm × 10 cm

Looking for similar products? Visit Product Comparison Guide

General description

Hybond™N+ is a high sensitivity positively charged nylon membrane for DNA and RNA blotting with nucleic acid binding capacity and high tensile strength.

Nucleic acid samples may be fixed by UV exposure or baking, although UV fixation gives better reproducibility. Hybond-N+ is not suitable for alkali blotting and fixation. All Hybond membranes are manufactured specifically for life science applications. Production runs are carefully controlled and the product exhaustively screened to ensure that only the most consistent product reaches the user. All Hybond membranes are identical on both sides. Supported nylon membranes have a high binding capacity for nucleic acid, in addition to high tensile strength. For applications requiring a high degree of sensitivity and/or reprobing these types of membranes are an ideal choice. Due to its high protein binding capacity, nylon requires extensive blocking prior to detection with antibodies to avoid high backgrounds. Consequently, nylon membranes are not recommended for use in Western blotting.

Application

Hybond™-N+ hybridization membranes are ideal to use with:

  • Positively charged nylon membrane with a binding capacity for nucleic acids up to 600 µg/cm² recommended for use with radioactive or nonradioactive chemiluminescence and chemifluorescence detection systems.
  • All membrane arrays with a high density.
  • Applications that need high sensitivity and/or probing.

Features and Benefits

  • On both sides, all Hybond™membranes are identicalIncompatible with alkali blotting and fixation.
  • To ensure that only the most consistent product reaches the user, production runs are carefully monitored, and the product is thoroughly screened.
  • Specially designed for life science applications.
  • In conventional southern blotting, produces excellent sensitivity.
  • To prevent high background sides, nylon needs to be extensively blocked before being detected with antibodies due to its high protein binding capacity.
  • Not suitable for western blotting.

Legal Information

Hybond is a registered trademark of Cytiva

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service