Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GERPN1234

Streptavidin-Alkaline

Cytiva RPN1234

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

packaging

pkg of 2 mL

manufacturer/tradename

Cytiva RPN1234

color

Colorless

storage temp.

2-8°C

General description

Streptavidin-Alkaline Phosphatase Conjugate.
Suitable for use in enhanced chemiluminescence (ECL).

application

Streptavidin-alkaline phosphatase is prepared using a single-step glutaraldehyde coupling reaction to link calf intestinal alkaline phosphatase to streptavidin. Excess coupling reagent is removed using extensive dialysis. The final product concentration is optimized to a standard titer as determined by its binding to biotinylated Protein. As a result the final Protein concentration may vary minimally between batches.

Features and Benefits

  • Streptavidin conjugated to reporter enzyme molecules for use in blotting and in situ hybridization applications.
  • High specificity reagents.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

pictograms

Exclamation mark

signalword

Warning

hcodes

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Using airbrushes to pattern reagents for microarrays and paper-fluidic devices.
Cassano CL, et al.
Microsystems & Nanoengineering, 3, 17055-17055 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service