Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GEPA22000

Sigma-Aldrich

Cy®2 bis-Reactive Dye Pack

Cytiva PA22000, pack of 5

Synonym(s):

Cy®2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

packaging

pack of 5

manufacturer/tradename

Cytiva PA22000

storage condition

protect from light

color

Yellow

storage temp.

2-8°C

General description

Cy2 bis-Reactive Dye Pack.

Application

Contains fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups. Each kit contains five vials of dye. Each vial is sufficient for labeling 1 mg of Protein or 30 nmol of amino groups.

Features and Benefits

  • Convenient way to label antibodies, Proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes.
  • For use in ELISA, immunoprecipitation, and blotting applications.
  • Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

CyDye:
Cy3B: This product is manufactured under an exclusive license from Carnegie Mellon University and is covered by US patent number 6,133,445 and equivalent patents and patent applications in other countries.
CyQ (or Cy5Q or Cy7Q): These products are covered under US patent number 6,828,116 and equivalent patents and patent applications in other countries in the name of Cytiva.
CytoCy5S: The use of this product in an NTR gene reporter assay is the subject of US patent number 7,579,140 in the name of Cytiva.
CypHer5E: This product or portions thereof is manufactured under license from Carnegie Mellon University and is covered by US patent number 7,615,646.
Cy and CyDye, are - trademarks of Cytiva or one of its subsidiaries.
The purchase of CyDye products includes a limited license to use the CyDye products for internal research and development but not for any commercial purposes. A license to use the Cy and CyDye trademarks for commercial purposes is subject to a separate license agreement with Cytiva. Commercial use shall include:
1. Sale, lease, license or other transfer of the material or any material derived or produced from it.
2. Sale, lease, license or other grant of rights to use this material or any material derived or produced from it.
3. Use of this material to perform services for a fee for third parties, including contract research and drug screening.
If you require a commercial license to use the Cy and CyDye trademarks please contact [email protected].

If you require a commercial license to use this material and do not have one, return this material unopened to Cytiva Bio-Sciences AB, Bjorkgatan 30, SE-751 84 Uppsala, Sweden and any money paid for the material will be refunded.
Cy is a registered trademark of Cytiva
CyDye is a trademark of Cytiva

pictograms

Exclamation mark

signalword

Danger

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service