Key Documents
GEHE99X1515
GE HE 99X max submarine unit
complete, basic, W 15 cm, L 10, 15, or 20 cm
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
packaging
pack of 1 ea
manufacturer/tradename
Cytiva Article No. 80-6061-57
Cytiva HE99X-15-1,5
General description
A versatile Max Submarine Unit ideal for the molecular biology lab, offers a variety of gel lengths and comb sizes so that you can run exactly the gel you need. Cast gels 15 cm wide × 10, 15, or 20 cm long, with one or two combs. Comb choices range from a preparative comb, for a 1 ml fragment purification sample, to a 30-well comb for high-throughput PCR screening. An adjustable comb back enables you to control the depth of sample wells. Run up to 60 samples at once and multichannelpipettor-compatible combs available.
Every design detail contributes to reliability and durability: seamless molded high impact plastic buffer tray; banana plugs safe from corrosion; a guard to protect the platinum electrodes; electrode carrier for simple electrode replacement; a rugged molded lid with safety guards to protect against electrical hazard; a large diameter bubble level and leveling feet; strain-relieved high voltage power cords; and red and black details on running trays and buffer chamber for orientation.
Durability and ease of use are engineered into the running trays: thick UVT acrylic; self-locating/centering guides to prevent tray floating or sliding; tray handles that align combs squarely at either end or the center of the gel; and flush tray ends for foam seal, or taping if you wish. Running trays seat snugly into the casting tray where the ends are sealed by closed-cell foam strips. No tape or loose parts required; the agarose stays where it belongs - on the running tray. For special non-submarine applications, you have the option of using the Max agarose dam casting feature.
Choose from one of the most recent versions:
Certificates of Analysis (COA)