Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE90100480

HiTrap® Chelating Sepharose FF

pkg of 5 × 5 mL, Cytiva, 90-1004-80, 90 μm avg. part. size, median particle size of the cumulative volume distribution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NB.21
Product GE90100480 is not currently sold in your country. Contact Technical Service

feature

median particle size of the cumulative volume distribution

packaging

pkg of 5 × 5 mL

manufacturer/tradename

90-1004-80
Cytiva

avg. part. size

90 μm

Legal Information

HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service