Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GE80-6501-04

2-D Fractionation Kit

Cytiva 80-6501-04, sufficient for 10 preparations

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for 10 preparations

feature

wetted part: no

manufacturer/tradename

Cytiva 80-6501-04

packaging

pkg of 10 samples

General description

The 2-D fractionation kit simplifies analysis of complex protein mixtures by reducing the amount and number of protein species loaded into the gel matrix.

Application

Fractionation makes it possible to isolate groups of Proteins, or fractions from the total proteome. This allows for improved resolution when an individual fraction is analyzed, provides less crowded 2-D maps, simplifies analysis and interpretation, and increases the chances of discovering novel Proteins of diagnostic or therapeutic interest.

The 2-D Fractionation Kit simplifies analysis of complex Protein mixtures by reducing the amount and number of Protein species loaded into the gel matrix.

Features and Benefits

  • Proven technology resolves complex Protein samples into six easily manageable fractions
  • High sample recovery with no loss of Protein
  • Produces more abundant spots.
  • Enhances the visibility and detection of low-abundance Proteins.
  • Can be scaled up for larger samples
  • Compatible with other downstream separation techniques, such as 2-D electrophoresis.

Storage and Stability

Lysis buffer: Once opened, s-re at 2-8 °C Fraction precipitant: S-re at ambient Precipitant: S-re at ambient Co-precipitant: S-re at ambient Wash buffer: Place at -20 °C for 1-2 hours before first use and thereafter s-re -20 °C Solubilizer: S-re powder at ambient Diluent: Once opened, s-re at 2-8 °C
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service