Key Documents
GE80648736
GE Ettan DALTsix gradient maker
volume 1000 mL
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
packaging
pack of 1 ea
manufacturer/tradename
Cytiva 80-6487-36
Cytiva Article No. 80-6487-36
volume
1000 mL
General description
The DALTsix Gradient Maker is designed to produce linear, convex and concave exponential gradients of solutions ranging in volume from 200 to 1000 ml. A sliding valve at the bottom quickly connects or disconnects the chambers.
Legal Information
Ettan is a trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)