Key Documents
GE80-6475-58
GE Ettan DALT low fluorescence glass plate set
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
packaging
pack of 1 ea
manufacturer/tradename
Cytiva 80-6475-58
Cytiva Article No. 80-6475-58
General description
Set contains:
1 x Borosilicate glass plate 27.6 cm x 21.7 cm x 3 mm
1 x Borosilicate glass plate 27.6 cm x 21.1 cm x 3 mm
2 x Vinyl spacer, 10 mm x 1 mm
The plates are for use of spot picker in combination with fluorocent dyes
Legal Information
Ettan is a trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This page describes two-dimensional separation of protein samples with Cytiva products.