Key Documents
GE80-6467-03
GE Ettan DALT blank cassette insert
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
packaging
pack of 1 ea
manufacturer/tradename
Cytiva 80-6467-03
Cytiva Article No. 80-6467-03
General description
When not casting cast a full set of gels use the blank cassette inserts with separator sheets between them to occupy the extra space. The Ettan DALTsix Gel Caster accommodates up to six 1-mm or 1.5-mm gel cassettes with separator sheets (0.5 mm) between them.
Legal Information
Ettan is a trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This protocol shows how to insert gels into Ettan™ DALT electrophoresis units.