Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE80-6418-77

MiniVe Vertical Electrophoresis System

AC/DC input, pack of

Synonym(s):

MiniVE Vertical Electrophoresis System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

description

Tank volume: 1.7 L with one module, 1.2 L with two modules

manufacturer/tradename

Cytiva 80641877

packaging

pack of

parameter

5-75 °C temp. range

Looking for similar products? Visit Product Comparison Guide

General description

The simple, modular design of miniVE vertical electrophoresis system consolidates all the small parts for electrophoresis and blotting into one convenient unit. Uniform gel temperature is maintained with the tank-style lower buffer chamber. A full tank stabilizes the temperature so that lanes are straight and bands are flat. The blot Module will accept two gel/membrane stacks simultaneously. The lower buffer chamber is designed to hold two Blot Modules, allowing four gels to be blotted at once

Features and Benefits

  • Load, clamp, and cast: The gel module is simultaneously a casting stand and an upper buffer chamber with integrated clamps. Simply assemble the gel sandwich of plates and spacers, clamp it onto the upper buffer chamber, then pivot the sealing gasket and cast the gel. After polymerization, simply release the bottom seal and drop the assembly into the tank for running. No extra parts are needed and no unclamping until the run is finished
  • Extra length and compatible dimensions: The miniVE handles gel cassettes up to 10 × 10.5 cm (W × H), which increases the separation length by more than 30% over conventional 10 × 8 cm gels. In addition, miniVE is compatible with the glass plates, combs, and spacers of SE 260 Mini-Vertical units. For gradient gels, mini-Vertical Gel Casters can produce up to four identical gels at one time.
  • The right power supplies: EPS 301 is the power supply of choice for the miniVE System. With ratings of 300 V and 400 mA, it is capable of both running and blotting gels. For faster transfers when blotting four gels at a time, use the EPS 2A200 for its higher current capability.

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service