Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE80-1255-53

ExcelGelprecast gels for SDS-PAGE

8-18 gel concentration, pack of 6 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

material

polyacrylamide
polyester film

form

solid

feature

wetted part: no

packaging

pack of 6 ea

manufacturer/tradename

Cytiva 80125553

parameter

15-200 kDa separation range

L × thickness

110 mm × 0.5 mm

color

Colorless

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Legal Information

ExcelGel is a trademark of Cytiva

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service