Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE29-0588-05

Antibody package, Protein A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.56

manufacturer/tradename

Cytiva

storage temp.

2-8°C

General description

Protein A package is recommended for the purification of human antibodies.

Contents
HiTrap Protein A HP (1 × 1 ml)
HiTrap Desalting (1 × 5 ml)

The Protein A package includes HiTrap Protein A HP column which is recommended for human antibody purification. A HiTrap Desalting column is also provided for quick pH neutralization of the eluted antibodies.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

pictograms

Flame

signalword

Warning

hcodes

Storage Class

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service