Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE29-0362-31

Superdex® Peptide PC 3.2/300

Cytiva 29-0362-31, column L × I.D. 30 cm × 3.2 mm, 13 μm avg. part. size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

borosilicate glass column
titanium frit

mol wt

100-7000 Da (peptides)

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 29-0362-31

availability

not available in USA, Canada and Puerto Rico

parameter

<50 mL sample volume
0.01-0.15 mL/min flow rate
2 Mpa (290 psi)

technique(s)

HPLC: suitable

bed volume

2.4 mL

column L × I.D.

30 cm × 3.2 mm

avg. part. size

13 μm

cleaning

1-14

working range

1-14

separation technique

size exclusion (SEC)

General description

Superdex Peptide PC 3.2/300 is a prepacked Precision Column for high-resolution gel filtration of peptides and other small biomolecules with molecular weights between 100 and 7000, in the micropreparative scale.Precision Column Holder enables use of precision columns with high-performance chromatography systems such as AKTA design.

Features and Benefits

  • Excellent choice for the polishing step in a purification procedure.
  • Precision columns (PC) for micropurification and analysis: sample volumes of 4 - 50 μL
  • Easy and predictable scale-up.
  • Excellent reproducibility and durability.
  • New hardware design for convenient direct connection to high performance chromatography systems such as ÄKTA design.

Suitability

Stable in all common buffers: 1 M acetic acid, 8 M urea, 6 M guanidine hydrochloride, 30% isopropanol, 70% ethanol, 1 M NaOH (for cleaning in place)
for high-resolution gel filtration of peptides and other small biomolecules

Storage and Stability

Store at 4 to 30 °C (20% ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Superdex is a registered trademark of Cytiva

pictograms

Flame

signalword

Warning

hcodes

Storage Class

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service