Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-9513-77

Tio2 Mag Sepharose

Cytiva 28-9513-77, pack of 4 × 500 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

TiO2

packaging

pack of 4 × 500 μL

manufacturer/tradename

Cytiva 28-9513-77

parameter

Room temperature temp. range

matrix

paramagnetic, spherical, highly cross-linked agarose particles

General description

TiO2 Mag Sepharose is magnetic beads designed for enrichment of phosphopeptides from tryptic digested protein samples.

Application

TiO2 Mag Sepharose magnetic beads are based on titanium dioxide (TiO2) chromatography and are designed to simplify capture and enrichment of phosphopeptides. TiO2 has high affinity for phosphopeptides and is an efficient method for enrichment of phosphopeptides from complex samples. TiO2 Mag Sepharose improves ease-of-use in sample preparation of phosphopeptides. The magnetic bead format has excellent properties for small-scale experiments. The high density of the beads allows rapid capture by magnetic devices while the visibility of the beads ensures reliable collection of all bound target peptides. Together with MagRack 6, a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel

Features and Benefits

  • Magnetic beads for selective capture and enrichment of phosphopeptides
  • Easy to use - visible and dense magnetic beads
  • Simple protocol with elution conditions optimized for MS analysis
  • Parallel handling of samples - six samples can be processed in less than one hour

Storage and Stability

Store at 4 to 8 °C. (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

pictograms

Flame

signalword

Warning

hcodes

Storage Class

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service