GE28-9480-20
Albumin IgG Depletion Spintrap™
Cytiva 28-9480-20, pack of 10 columns
Synonym(s):
Albumin Depletion Column, Albumin Removal Spintrap, IgG Depletion Spintrap
About This Item
Recommended Products
ligand
recombinant protein G fragment and recombinant protein binding HSA
feature
wetted part: no
packaging
pack of 10 columns
manufacturer/tradename
Cytiva 28-9480-20
storage condition
(20% Ehtanol)
(20% Ethanol)
parameter
<4 mL/min flow rate
bed volume
370 μL
avg. part. size
34 μm
cleaning
2-9
working range
3-9
storage temp.
2-8°C
General description
Features and Benefits
- High depletion capacity, removes > 95% albumin and > 90% IgG,with high reproducibility
- Removal of albumin and IgG allows a higher load of less abundant proteins to be included in the analysis and detection of more proteins
- Simple and fast procedure
- HiTrap Albumin and IgG Depletion provides rapid and easy processing in a convenient format for handling larger sample volumes (~150 μl) using a liquid chromatography system or manually using a syringe
- Albumin and IgG Depletion SpinTrap handles smaller sample volumes (~50 μl) using a tabletop centrifuge
Storage and Stability
Analysis Note
Legal Information
signalword
Warning
hcodes
Storage Class
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This page shows how to purify or remove albumin and IgG with Albumin & IgG Depletion Sepharose High Performance from Cytiva.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service