GE28-9355-99
Streptactin Sepharose™ High Performance
Cytiva 28-9355-99, pack of 10 mL
About This Item
Recommended Products
ligand
StrepTactin
packaging
pack of 10 mL
manufacturer/tradename
Cytiva 28-9355-99
storage condition
(20% Ehtanol)
matrix
highly cross-linked 6% agarose
avg. part. size
34 μm
working range
7
capacity
~6 mg binding capacity (Strep II-tagged protein/ml medium)
shipped in
wet ice
storage temp.
2-8°C
General description
Features and Benefits
- Highly pure Strep II-tagged recombinant proteins eluted in concentrated form and small volumes
- Physiological conditions and mild elution preserve target protein activity
- Fast and easy regeneration with 0.5 M NaOH
- Compatible with a wide range of reducing agents, detergents, denaturants and other additives
Storage and Stability
Analysis Note
Legal Information
signalword
Warning
hcodes
Storage Class
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
This page shows how to purify Strep-tag II recombinant proteins using StrepTactin Sepharose® High Performance from Cytiva.
This page shows how to purify Strep-tag II recombinant proteins using StrepTrap HP 1 mL and 5 mL from Cytiva.
This page describes efficient column packing and preparation for affinity chromatography of tagged proteins using Cytiva products.
This page covers detailed information on cleaning procedures and recommended flow for column cleaning.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service