Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28931733

HiTrap® PlasmidSelect Xtra

pkg of 5 × 5 mL, Cytiva, 28-9317-33

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NB.21
Product GE28931733 is not currently sold in your country. Contact Technical Service

packaging

pkg of 5 × 5 mL

manufacturer/tradename

28-9317-33
Cytiva

Legal Information

HiTrap is a registered trademark of Cytiva

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service