Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GE28-9269-76

HiScreen MabSelect Xtra

Cytiva 28-9269-76

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

recombinant protein A (E. coli)

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 28-9269-76

storage condition

(20% Ehtanol)
(20% Ethanol)

parameter

44 psi

bed size

7.7 mm × 100 mm

bed volume

4.7 mL

column I.D.

7.7 mm

matrix

highly cross-linked agarose

avg. part. size

75 (d₅₀v is the median particle size of the cumulative volume distribution)

cleaning in place

2-12

working range

3-10

capacity

≥30 mg binding capacity (humanlgG/ml at 2.4 min residence time)

suitability

suitable for bioprocess medium

storage temp.

2-8°C

Related Categories

General description

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Features and Benefits

  • The increased capacity of MabSelect Xtra enables purification of MAbs from high-level expression feedstock.
  • Prepacked, ready-to-use columns for convenient process development.
  • 1NA cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 2NA cm bed height.
  • Small bed volume gives fast results and minimal sample/buffer consumption.*Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiScreen is a trademark of Cytiva
MabSelect Xtra is a trademark of Cytiva

pictograms

Flame

signalword

Warning

hcodes

Storage Class

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service