GE28-9075-47
StrepTrap™ High Performance prepacked chromatography column
pkg of 1 × 5 mL, prepacked chromatography column
Synonym(s):
StrepTrap™ High Performance
About This Item
Recommended Products
ligand
StrepTactin
packaging
pkg of 1 × 5 mL
manufacturer/tradename
Cytiva 28-9075-47
storage condition
(20% Ehtanol)
(20% Ethanol)
parameter
<20 mL/min flow rate
42 psi (H2O at room temperature.)
bed size
16 mm × 25 mm
bed volume
5 mL
column I.D.
16 mm
matrix
highly cross-linked 6% agarose
avg. part. size
34 μm
working range
7
capacity
~6 mg binding capacity (Strep II-tagged protein/ml medium)(H2O at room temperature.)
suitability
suitable for chromatography
storage temp.
2-8°C
General description
Application
Features and Benefits
- Highly pure Strep-tag II recombinant proteins eluted in concentrated form and small volumes.
- Physiological conditions and mild elution preserves the activity of the target protein.
- Convenient, time-saving operation and reproducible results.
Storage and Stability
Analysis Note
Other Notes
Legal Information
signalword
Warning
hcodes
Storage Class
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
This page describes manual and automated purification of histidine-tagged recombinant proteins using Cytiva products
Protocols
This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service