Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-9042-63

illustra Blood Mini Spin (10)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NA.52

manufacturer/tradename

Cytiva

General description

The purification process involves minimal shearing resulting in the production of 5 to 10 μg of good quality, intact genomic DNA from a 200 μl sample. Input sample volumes can vary from 50 μl to 1 ml. The method uses a simple genomic DNA purification protocol that uses chaotropic agents to extract DNA from blood cells, denature protein components, and promote the selective binding of DNA to a column-based, novel silica membrane. The elution volume can be modified as necessary to obtain the appropriate genomic DNA concentration for most downstream molecular biological applications. The kit contains spin columns prepacked with a novel silica membrane, lyophilized proteinase K powder, lysis solution, wash and elution buffers, microcentrifuge collection tubes, a full protocol booklet, and a detachable, quick reference protocol card.

Features and Benefits

Demonstrated application to various types of whole blood including human, horse, rabbit, rat, and mouse.
For the rapid and reproducible isolation of high-quality genomic DNA from whole blood, buffy coat, bone marrow, and nucleated red blood cells.
High-quality genomic DNA with a greater proportion of higher molecular weight DNA and less shearing.
Increased ease of use with reduced pipetting volume changes, one centrifugation setting, and color-coded kit components
Verified in several downstream applications including real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digests.

Legal Information

illustra is a trademark of Cytiva

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service