Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-4065-41

2′-Deoxyuridine 5′-Triphosphate Solution, 100 mM

Cytiva 28-4065-41, pack of 25 μmol

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$64.40

$64.40


Usually ships in 2 weeks.


Select a Size

Change View
50 G
$64.40

About This Item

UNSPSC Code:
41106305
NACRES:
NA.52

$64.40


Usually ships in 2 weeks.

form

liquid

packaging

pack of 25 μmol

manufacturer/tradename

Cytiva 28-4065-41

concentration

100 mM

color

Colorless

shipped in

dry ice

storage temp.

−20°C

General description

2′-Deoxyuridine 5′-Triphosphate Solution, 100 mM.

application

By incorporating recent advances in manufacturing technology, Cytiva now offers illustra Solution dNTPs: the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.

Features and Benefits

  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR* and sequencing
  • Custom formulation, testing, packaging, and labeling are available

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

illustra is a trademark of Cytiva

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service