Key Documents
GE28-4064-19
Sigma-Aldrich
Tricorn 10/600 Column
About This Item
UNSPSC Code:
12352200
NACRES:
SB.52
technique(s)
LPLC: suitable
General description
Tricorn 5 and 10 Coarse Filter Kit is used for large volume/repeated loading of clarified sample to reduce the risk of clogging in combination with capture media. Both filter and coarse filter kits include top and bottom adapters, bottom filters, and EPDM O-rings, five of each.
Tricorn Packing Equipment includes the Packing Equipment 5/50 (5-mm connector and 50-mm glass tube) or 10/100 (10-mm connector and 100-mm glass tube), EPDM O-rings and bottom unit, cap, and stop plug.
Features and Benefits
Broad range of column dimensions suitable for many different purification needs.
Glass tubes coated with plastic film prevent harm if the column is dropped or operated outside its specification.
Locking ring prevents accidental compression of media bed.
Separate packing connector allows simple column packing for a broad range of media.
The modern integrated, snap-on design results in a column that is easy to assembly, operate, and maintain.
Tricorn empty columns are designed for high performance separations on small and medium scale. Pressure stability up to 100 bar and durable column material ensure reliable and reproducible operation as well as compatibility with the majority of the commercial lab media.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
This page covers the standard ÄKTAdesign configurations for simple IEX chromatography.
This page shows volatile and non-volatile buffer suggestions for anion and cation exchange chromatography.
This page covers practical problems that may lead to a non-ideal IEX separation and their solutions.
This page covers detailed aspects of each step in an IEX separation to improve resolution and overall performance.
Protocols
This page covers the use of Sepharose High Performance media for purification of proteins, peptides or oligonucleotides, when to use them, and with which systems.
This page covers the use of Sepharose Fast Flow for purification of proteins.
Column Packing and Preparation for Size Exclusion Chromatography
This page shows how to perform column packing and preparation for ion exchange chromatography and chromatafocusing when using Tricorn or XK columns available from Cytiva.
See All