Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-4013-51

His GraviTrap Kit

Cytiva 28-4013-51, kit of one pack of His buffer, pack of 2 × 10 columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

manufacturer/tradename

Cytiva 28-4013-51

packaging

kit of one pack of His buffer, pack of 2 × 10 columns

storage condition

(20% Ethanol)

bed volume

1 mL

capacity

40 mg binding capacity (histidine-tagged protein/column)(Protein binding capacity is protein-to-protein dependent.)

storage temp.

2-8°C

General description

His Buffer Kit contains carefully prepared buffer concentrates for binding, washing, and elution of histidine-tagged proteins according to recommended protocols. Moreover, method for optimization of imidazole concentration in the binding buffer is described.

His GraviTrap is a prepacked, single-use column for purification of histidine-tagged proteins by immobilized metal affinity chromatography (IMAC). Column gives fast and simple gravity-flow purifications without any need for a purification.

The ready to use column is prepacked with Ni Sepharose 6 Fast Flow, which has a high protein binding capacity, low nickel-ion leakage, and is compatible with denaturing and reducing agents, as well a wide range of additives.

His GraviTrap columns are delivered in a package that can be converted into a column stand to simplify purification. LabMate PD-10 Buffer Reservoir can be connected to the columns for convenient handling of sample volumes above 10 mL.
The Buffer kit can also be separately purchased:.
The His GraviTrap can also be separately purchased:
.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

GraviTrap is a trademark of Cytiva

related product

Product No.
Description
Pricing

pictograms

Health hazard

signalword

Danger

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page shows how to purify histidine-tagged recombinant proteins by gravity flow using His GraviTrap and His GraviTrap Kit from Cytiva.

Ni Sepharose 6 Fast Flow purifies histidine-tagged proteins efficiently, offering high cross-linked agarose beads with Ni2+ ions.

This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service