Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE27-4810-01

Pezz 18 Expresion Vector

Cytiva 27-4810-01, pack of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000

packaging

pack of 1 ea

manufacturer/tradename

Cytiva 27-4810-01

shipped in

dry ice

storage temp.

−20°C

General description

pEZZ 18 Protein A Gene Fusion Vector.

Application

The phagemid pEZZ 18 contains the Protein A signal sequence and two synthetic “Z” domains based on the “B” IgG binding domain of Protein A. Proteins are expressed as fusions with the “ZZ” peptide and secreted into the aqueous culture medium under the direction of the Protein A signal sequence. They are easily purified using IgG Sepharose 6 Fast FLow to which the “ZZ” domain binds tightly. Because of its folding properties, the 14 kDa “ZZ” peptide has little effect on folding of the fusion partner into a native conformation.

Expression is controlled by the lacUV5 and Protein A promoters and is not inducible. Elements of the Protein A gene provide the ATG and ribosome-binding sites. Stop codons must be provided by the insert.

M13 Universal Sequencing Primer can be used for both double-stranded and single-stranded sequencing. A protocol for production of single-stranded DNA is provided with the vector.

Inserts containing a stop codon will yield white colonies when grown on media containing X-gal.

Host(s). E. coli strains carrying a lac deletion but capable of a-complementation of lacZ′.

Features and Benefits

  • For rapid expression of secreted fusion Proteins and their one-step purification using IgG Sepharose 6 Fast FLow

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service