Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE27-2081-01

2′,3′-Dideoxythymidine 5′-Triphosphate, 100 mM solution (ddTTP)

Cytiva 27-2081-01

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106305
NACRES:
NA.52

form

liquid

packaging

pkg of 40 μL

manufacturer/tradename

Cytiva 27-2081-01

concentration

100 mM

color

Colorless

shipped in

dry ice

storage temp.

−20°C

General description

2′,3′-Dideoxythymidine 5′-Triphosphate, 100 mM solution (ddTTP).

Features and Benefits

  • Easy-to-use, time-saving solutions for use in any application requiring ddNTPs of high purity, such as dideoxy sequencing and genotyping reactions.
  • High purity: ? 98% triphosphate.
  • Each lot of ddNTPs is functionally tested to meet or exceed criteria for high-quality sequencing with Thermo Sequenase DNA Polymerase.
  • Available either as individual products or in a convenient set containing ddATP, ddCTP, ddGTP, and ddTTP in separate tubes.
  • Supplied in purified water as 5 mM and 100 mM solutions.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service