GE27-0849-01
Factor Xa
Cytiva 27-0849-01, pack of 400 units
About This Item
Recommended Products
packaging
pack of 400 units
manufacturer/tradename
Cytiva 27-0849-01
shipped in
dry ice
storage temp.
−20°C
General description
Application
Features and Benefits
- Enables the site-specific cleavage of fusion Proteins containing an accessible Factor Xa protease recognition sequence.
- Can be used either following affinity purification or while fusion Proteins are bound to Glutathione Sepharose™ 4B, Glutathione Sepharose™ High Performance, or Glutathione Sepharose™ 4 Fast Flow.
Storage and Stability
Analysis Note
Legal Information
Storage Class
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service