Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE25-8008-60

Sigma-Aldrich

CyDye DIGE Cy®2 Minimal 10 nmol

Cytiva 25-8008-60

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

form

solid

feature

wetted part: no

packaging

pkg of 10 nmol

manufacturer/tradename

Cytiva 25-8008-60

storage condition

protect from light

color

Yellow

solubility

cold water: soluble (Soluble in cold water and hot water)

shipped in

dry ice

storage temp.

−20°C

General description

CyDye DIGE Fluor, Cy2 minimal dye. CyDye DIGE Fluor minimal dyes and saturation dyes are size- and charge-matched fluorescent dyes specifically designed for detecting protein abundance differences in 2-D Fluorescence Difference Gel Electrophoresis using Ettan DIGE system.

Application

CyDye DIGE fluors are available as minimal and saturation labeling dyes. The minimal dyes are intended for general 2-D application use where sufficient amounts of sample are available. The saturation dyes from the Scarce Sample Labeling Kit are designed to be used for applications where only small amounts of sample are available, for example in Laser Capture Microdissection. In total, three minimal dyes and two saturation dyes are available.

CyDye DIGE fluors are exceptional dyes for multicolor analysis, offering bright and intense colors with narrow excitation and emission bands. The fluors are spectrally distinct, making them highly suitable for multicolor detection. CyDye DIGE fluors utilize these benefits but are also size- and charge-matched specifically for 2-D DIGE using Ettan DIGE system.

CyDye DIGE Fluor Minimal Dyes
Protein samples and the internal standard are each labeled with one CyDye DIGE Fluor minimal dye. These labeled samples are then combined, run on an isoelectric focusing gel in the first dimension, and separated by SDS-PAGE in the second dimension. Electrophoresis is simplified with Ettan IPGphor 3, or Multiphor II with Immobiline® DryStrip gels in the first dimension and Ettan DALTtwelve or Ettan DALTsix electrophoresis systems in the second dimension.

The ability to multiplex different CyDye DIGE Fluor minimal dye-labeled samples on the same gel means that the different samples will be subjected to exactly the same first- and second-dimension running conditions. Consequently, the same Protein labeled with any of the CyDye DIGE Fluor minimal dyes and separated on the same gel will migrate to the same position on the 2-D gel and overlay. This limits experimental variation and ensures accurate within-gel matching.

Features and Benefits

  • AlLow detection of up to three prelabeled Protein samples and standards on the same 2-D electrophoresis gel.
  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel.
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique.
  • Minimal loss of signal during labeling, separation, and scanning.
  • No change in signal over wide pH range used during first-dimension (IEF) separation.
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy.

Packaging

10 nmol (2 x 5 nmol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

CyDye:
Cy3B: This product is manufactured under an exclusive license from Carnegie Mellon University and is covered by US patent number 6,133,445 and equivalent patents and patent applications in other countries.
CyQ (or Cy5Q or Cy7Q): These products are covered under US patent number 6,828,116 and equivalent patents and patent applications in other countries in the name of Cytiva.
CytoCy5S: The use of this product in an NTR gene reporter assay is the subject of US patent number 7,579,140 in the name of Cytiva.
CypHer5E: This product or portions thereof is manufactured under license from Carnegie Mellon University and is covered by US patent number 7,615,646.
Cy and CyDye, are - trademarks of Cytiva or one of its subsidiaries.
The purchase of CyDye products includes a limited license to use the CyDye products for internal research and development but not for any commercial purposes. A license to use the Cy and CyDye trademarks for commercial purposes is subject to a separate license agreement with Cytiva. Commercial use shall include:
1. Sale, lease, license or other transfer of the material or any material derived or produced from it.
2. Sale, lease, license or other grant of rights to use this material or any material derived or produced from it.
3. Use of this material to perform services for a fee for third parties, including contract research and drug screening.
If you require a commercial license to use the Cy and CyDye trademarks please contact [email protected].

If you require a commercial license to use this material and do not have one, return this material unopened to Cytiva Bio-Sciences AB, Bjorkgatan 30, SE-751 84 Uppsala, Sweden and any money paid for the material will be refunded.
Cy is a registered trademark of Cytiva
CyDye is a trademark of Cytiva
Ettan is a trademark of Cytiva
Immobiline is a registered trademark of Cytiva

pictograms

Exclamation mark

signalword

Danger

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Troubleshooting table for 2D DIGE results lists problems, causes, and prevention methods for future experiments.

Protocols

This page shows how to perform a proteomic analysis of membrane proteins to identify and quantitate all proteins expressed in a cell or a tissue with products from Cytiva.

This page describes two-dimensional separation of protein samples with Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service