Key Documents
GE19-5208-01
Sigma-Aldrich
RC 26 Packing Reservoir
About This Item
UNSPSC Code:
41115716
NACRES:
SB.52
technique(s)
LPLC: suitable
General description
RC packing reservoirs, which are available for all three C column types, are quick and easy to assemble. Curved walls minimize the separation of matrix beads by size during packing to produce homogeneous columns. Once the matrix is in the column, the RC reservoir can be connected to a pump or Mariotte flask to give optimal flow rates for column packing.
Features and Benefits
Packing reservoirs for each column diameter optimize packing conditions.
Choose from one of the most recent versions:
Certificates of Analysis (COA)