Key Documents
GE18114710
Fingertight Narrow Head Connector
O.D. (1/16 in.), ÄKTA® Systems, Cytiva 18114710
About This Item
UNSPSC Code:
41116500
NACRES:
NB.43
material
PEEK
feature
Connections: Fingertight 1/16"
Wetted Material: Polyetheretherketone (PEEK)
ÄKTA® Systems
manufacturer/tradename
Cytiva 18114710
O.D.
(1/16 in.)
1.6 mm
compatibility
For use with: Tubing O.D. 1/16"
Legal Information
ÄKTA is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)