Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-6002-50

ETTAN DALT Buffer Kit

Cytiva 17600250, liquid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

sterility

non-sterile

form

liquid

feature

wetted part: no

manufacturer/tradename

Cytiva 17600250

storage temp.

2-8°C

General description

Dalt gel 12.5 (26 x 20 cm) is a precast polyacrylamide gel bound to a plastic support film, which provides ease of handling and dimensional stability. The gels are barcoded for convenient sample and batch tracking. Dalt gel 12.5 is used together with dalt buffer kit, which comprises concentrated buffers for running the gel, gel buffer for inserting the gel into dalt precast gel cassette, and sealing solution for application of the ipg strip. The buffer in the gel is part of a buffer system that gives longer shelf-life. Precast gel cassettes improve handling efficiency and throughput. Gels can be quickly loaded and unloaded with minimal hands-on time

Features and Benefits

DALT Buffer Kit for 12 Precast Gels

  • Highest capacity electrophoresis unit with the largest precast gels available (26 u 20 cm)
  • Stable, discontinuous buffer system in a precast, homogeneous gel features high batch-to-batch reproducibility
  • Simple-to-use gel cassettes for gel loading, unloading, and storage
  • Programmable power supply and temperature control ensure high reproducibility among runs.
  • System design minimizes buffer consumption.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This protocol shows how to insert gels into Ettan™ DALT electrophoresis units.

Related Content

This page shows how to apply equilibrated Immobiline DryStrip gels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service