Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17545109

HiTrap® Capto Phenyl (High Sub)

pkg of 5 × 5 mL, Cytiva, 17-5451-09, 75 μm avg. part. size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NB.21
Product GE17545109 is not currently sold in your country. Contact Technical Service

packaging

pkg of 5 × 5 mL

manufacturer/tradename

17-5451-09
Cytiva

avg. part. size

75 μm

General description

Capto Phenyl (High sub) and Capto Butyl are hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification

Legal Information

Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service