Recommended Products
shelf life
Estimated shelf life is 5 years from manufacture date.
manufacturer/tradename
Cytiva
matrix
highly cross-linked agarose
particle size
75 μm
cleaning
2-14
working range
3-12
suitability
suitable for bioprocess medium
General description
- Multimodal anion exchanger for post-protein A purification of monoclonal antibodies.
- High capacity and productivity in flow-through mode
- Removes contaminants to formulation levels.
- Wide operational window of pH and conductivity.
- Two-step chromatographic process with protein A affinity, saves time and cuts operating costs.
Capto adhere is a multimodal BioProcess medium designed for post-protein A purification of monoclonal antibodies (MAbs) at process scale. It is composed of a rigid, high-flow agarose matrix and a novel multimodal ligand. The strong multimodal ligand gives a different selectivity compared to traditional ion exchangers. Capto adhere can remove key contaminants such as DNA, host cell proteins (HCP), leached protein A, dimers and larger aggregates, and viruses in a single step. Together with a Protein A capture step, like MabSelect SuRe, Capto adhere allows the design of a two-step chromatographic process for Mabs.
For optimal performance of Capto adhere, screening/optimization of loading conditions is recommended, preferably using modern high-throughput process development tools, such as PreDictor 96-well filter plates.
Storage and Stability
Legal Information
signalword
Warning
hcodes
Storage Class
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service