Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-5176-01

Superdex® Peptide 10/300 GL

Cytiva 17-5176-01, column L × I.D. 30 cm × 10 mm, 13 μm avg. part. size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

borosilicate glass column
polypropylene filter

mol wt

100-7000 Da

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 17-5176-01

parameter

0.15-0.6 mL/min flow rate
1.8 Mpa (261 psi)
25-500 mL sample volume

bed size

10 mm × 310 mm

bed volume

24 mL

column L × I.D.

30 cm × 10 mm

matrix

dextran composite

avg. part. size

13 μm

cleaning

1-14

working range

1-14

separation technique

size exclusion (SEC)

General description

Superdex® Peptide 10/300 GL is a prepacked Tricorn high-performance column for high-resolution gel filtration of peptides and other small biomolecules with molecular weights between 100 and 7000.

Superdex® prepacked columns are designed for high-performance, laboratory-scale separations of proteins, peptides, and other biomolecules according to size. High performance gel filtration offers fast separations with high resolution for a variety of applications including protein purification, studies of complex formation, and screening of uncharacterized samples.

Features and Benefits

  • High-resolution separations of proteins, peptides, and other biomolecules according to size.
  • Well suited for the polishing step in a purification procedure.
  • Easy and predictable scale-up.
  • Excellent reproducibility and durability.

Suitability

Stable in all common buffers: 1 M acetic acid, 8 M urea, 6 M guanidine hydrochloride, 30% isopropanol, 70% ethanol, 1 M NaOH (for cleaning in place)
for high-resolution gel filtration of peptides and other small biomolecules

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Superdex is a registered trademark of Cytiva

pictograms

Flame

signalword

Warning

hcodes

Storage Class

"" - ""


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service