GE17-5174-01
Superdex® 75 10/300 GL
Cytiva 17-5174-01, column L × I.D. 30 cm × 10 mm, 13 μm avg. part. size
About This Item
Recommended Products
material
bed support (polyamide)
glass column
polypropylene fiber (reinforced glass)
product line
Superdex®
mol wt
3000-70000 Da (globular proteins)
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 17-5174-01
parameter
0.5 ml/min - 1 ml/min flow rate
1.8 Mpa (261 psi)
25-500 μL sample volume
bed volume
24 mL
column L × I.D.
30 cm × 10 mm
matrix
agarose-dextran composite
polymeric
avg. part. size
13 μm
cleaning
1-14
working range
3-12
separation technique
size exclusion (SEC)
General description
Superdex® prepacked columns are designed for high-performance, laboratory-scale separations of proteins, peptides, and other biomolecules according to size. High performance gel filtration offers fast separations with high resolution for a variety of applications including protein purification, studies of complex formation, and screening of uncharacterized samples.
Superdex® 75 5/150 GL and Superdex® 200 5/150 GL have small bed volumes, which is useful for a range of applications such as screening solubilization conditions for membrane proteins, the study of protein-protein interactions, and rapid purity check of proteins. These columns are an excellent choice when time and sample/buffer consumption is more important than the highest resolution since less buffer and sample volumes are required.
Superdex® columns 10/300 GL and 5/150 GL are prepacked Tricorn columns, see Tricorn Column Family excellent reproducibility and durability in purification using a chromatography system.
Features and Benefits
- High-resolution separations of proteins, peptides, and other biomolecules according to size.
- Well suited for the polishing step in a purification procedure.
- Separations in two ranges from Mr 3000 up to 600 000 (globular proteins).
- Easy and predictable scale-up.
- Excellent reproducibility and durability.
- For rapid protein homogeneity analysis or purity check use the 5/150 GL short columns.
Suitability
Storage and Stability
Analysis Note
Legal Information
signalword
Warning
hcodes
Storage Class
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service