Key Documents
GE17-1455-01
Precision Column Holder
Cytiva 17-1455-01
About This Item
UNSPSC Code:
23151816
NACRES:
NA.56
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 17-1455-01
General description
Adapts PC columns designed for SMART System and for use with other high performance chromatography systems including AKTAdesign systems.
Features and Benefits
  • Precision column holder contains two holders, one for 3 to 5 cm columns and one for 30 cm columns.
  • Low dead volume fittings maintain resolution.
  • Includes all necessary tubing fittings and instructions.
Storage Class
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)