Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-1330-01

Bind-silane

Cytiva 17-1330-01, pack of 25 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

form

liquid

feature

wetted part: no

packaging

pack of 25 mL

manufacturer/tradename

Cytiva 17-1330-01

color

Pale color

solubility

acetone: soluble
diethyl ether: soluble
methanol: soluble

storage temp.

room temp

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

pictograms

Exclamation mark

signalword

Warning

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service