GE17-1325-01
Glycerol 87%
Cytiva 17-1325-01, pack of 1000 mL
About This Item
Recommended Products
form
liquid
feature
wetted part: no
packaging
pack of 1000 mL
manufacturer/tradename
Cytiva 17-1325-01
color
Colorless
solubility
cold water: soluble (Soluble in cold water and hot water)
storage temp.
room temp
Storage and Stability
Analysis Note
Storage Class
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
This page describes typical rehydration solutions for use in gel electrophoresis with Cytiva products.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service