Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-1311-01

Ammonium Persulphate

Cytiva 17-1311-01, pack of 25 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

form

solid

feature

wetted part: no

packaging

pack of 25 g

manufacturer/tradename

Cytiva 17-1311-01

storage condition

protect from light

color

White

solubility

cold water: soluble (Soluble in cold water and hot water)

General description

PlusOne Ammonium Persulfate. The high quality of PlusOne Ammonium Persulfate supports efficient polymerization at concentrations as Low as 1 mM.

Application

Ammonium persulfate is the initiator of choice for the polymerization of acrylamide.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 15 to 25 °C. (drt)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Pictograms

Exclamation mark

signalword

Danger

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page describes typical rehydration solutions for use in gel electrophoresis with Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service