Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-1310-01

ReadySol IEF, 40%

Cytiva 17-1310-01, pack of 1000 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NB.22

form

liquid

feature

wetted part: no

packaging

pack of 1000 mL

manufacturer/tradename

Cytiva 17-1310-01

storage condition

protect from light

color

Colorless

solubility

cold water: soluble (Soluble in cold water and hot water)

storage temp.

2-8°C

General description

PlusOne Acrylamide IEF is outstanding for use in isoelectric focusing and DNA sequencing. In addition to the absence of polymerization interfering impurities, Acrylamide IEF is essentially free of charged acrylamide derivatives (< 20 ppm) for very stable pH gradients.

PlusOne Acrylamide is available as a powder, a solution, and premixed solutions (ReadySol). Dry powder is the most stable form of acrylamide with a shelf life of more than three years.

Features and Benefits

Solutions of acrylamide are convenient, virtually eliminating the risk of acrylamide dust in the air. ReadySol, a solution of acrylamide and bisacrylamide, is convenient and fast with minimal preparation and handling. ReadySol ensures a reproducible solution composition.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 10 to 30 °C ()

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

pictograms

Exclamation mark

signalword

Danger

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service