GE17-1304-02
Methylenebis Acrylamide
Cytiva 17-1304-02, pack of 100 g
About This Item
Recommended Products
form
solid
feature
wetted part: no
packaging
pack of 100 g
manufacturer/tradename
Cytiva 17-1304-02
color
White
pH range
5.5-7.0
solubility
acetone: soluble (Slightly soluble in acetone)
cold water: soluble (Soluble in cold water and hot water)
storage temp.
2-8°C
General description
Analysis Note
signalword
Warning
hcodes
Storage Class
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This page describes typical rehydration solutions for use in gel electrophoresis with Cytiva products.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service