Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-1302-02

Acrylamide Page

Cytiva 17-1302-02, pack of 1000 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

form

solid

feature

wetted part: no

packaging

pack of 1000 g

manufacturer/tradename

Cytiva 17-1302-02

storage condition

room temperature

color

White

pH range

>5.0(in 5% aq. soln.)

solubility

acetone: soluble
cold water: soluble (Soluble in cold water and hot water)
methanol: soluble

General description

PlusOne Acrylamide PAGE contains extremely low concentrations of impurities that can interfere with the polymerization reaction. It is especially suitable for SDS and native PAGE techniques.

Application

PlusOne Acrylamide is available as a powder, a solution, and premixed solutions (ReadySol). Dry powder is the most stable form of acrylamide with a shelf life of more than three years.

Features and Benefits

Solution
Solutions of acrylamide are convenient, virtually eliminating the risk of acrylamide dust in the air.

ReadySol
ReadySol, a solution of acrylamide and bisacrylamide, is convenient and fast with minimal preparation and handling. ReadySol ensures a reproducible solution composition.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

signalword

Danger

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

280.4 °F - closed cup

flash_point_c

138 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page describes typical rehydration solutions for use in gel electrophoresis with Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service